missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(Boc-aminooxy)acetic acid, 98%

Manufacturer:  Alfa Aesar™ H6287906

 View more versions of this product

Catalog No. AAH6287906


 

Description

Description

Applications
Bis(Boc-aminooxy)acetic acid employed as a protecting reagent and intermediate pharmaceuticals and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

Bis(Boc-aminooxy)acetic acid
98%
[(CH3)3COCONHO]2CHCOOH
White
5g
CC(C)(C)OC(=O)N(C(=O)OC(C)(C)C)OCC(=O)O
291.3
322.32
293302-31-5
C12H21NO7
MFCD06410966
Bis-Boc-Aoa
PXAKWBDTOCTFOJ-UHFFFAOYSA-N
2-[bis[(2-methylpropan-2-yl)oxycarbonyl]amino]oxyacetic acid
2755975
Powder

Chemical Identifiers

293302-31-5
291.3
PXAKWBDTOCTFOJ-UHFFFAOYSA-N
2755975
C12H21NO7
MFCD06410966
Bis-Boc-Aoa
SDS Documents