missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(benzonitrile)palladium(II) chloride, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 207900100

 View more versions of this product

Catalog No. AC207900100



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Bis(benzonitrile)palladium(II) chloride
99.0
120.0°C
99.0% min.
97+%
C14H10Cl2N2Pd
MFCD00013123
01,56; 05,31; 06,45; 09,44; 10,31; 13,34
Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, toluene, chloroform
Cl[Pd++](Cl)([N]#CC1=CC=CC=C1)[N]#CC1=CC=CC=C1
383.57
383.55
Crystalline Powder
14220-64-5
100.0
Orange to Brown
Authentic
Glass bottle
(C6H5CN)2PdCl2
10 g
bis benzonitrile palladium chloride, bis benzonitrile palladium ii chloride, trans-bis benzonitrile palladium ii chloride, bis benzonitrile palladium ii chloride, 500mg, palladium 2+ bis benzonitrile dichloride
QQPSWGUJSPTOSH-UHFFFAOYSA-L
benzonitrile;palladium(2+);dichloride
10271322
97+%

Chemical Identifiers

14220-64-5
383.57
QQPSWGUJSPTOSH-UHFFFAOYSA-L
10271322
Cl[Pd++](Cl)([N]#CC1=CC=CC=C1)[N]#CC1=CC=CC=C1
C14H10Cl2N2Pd
MFCD00013123
bis benzonitrile palladium chloride, bis benzonitrile palladium ii chloride, trans-bis benzonitrile palladium ii chloride, bis benzonitrile palladium ii chloride, 500mg, palladium 2+ bis benzonitrile dichloride
benzonitrile;palladium(2+);dichloride
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

EINECSNumber : 238-085-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only