Please login to your online account to display your discounted pricing

Bis(benzonitrile)palladium(II) chloride, 99+%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  207900010

 View more versions of this product

Catalog No. AC207900010

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Bis(benzonitrile)palladium(II) chloride
CAS 14220-64-5
CAS Min % 99.0
Elemental Analysis 99.0% min.
Health Hazard 1 GHS Signal Word: Warning
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 99+%
Molecular Formula C14H10Cl2N2Pd
Linear Formula (C6H5CN)2PdCl2
Formula Weight 383.55
MDL Number MFCD00013123
Color Orange to brown
Fieser 01,56; 05,31; 06,45; 09,44; 10,31; 13,34
Quantity 1g

Specifications

Chemical Name or Material Bis(benzonitrile)palladium(II) chloride
CAS 14220-64-5
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Molecular Formula C14H10Cl2N2Pd
Linear Formula (C6H5CN)2PdCl2
Formula Weight 383.55
MDL Number MFCD00013123
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.

SDS & Safety Documents