missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis[alpha,alpha-bis(trifluoromethyl)benzenemethanolato]diphenylsulfur, TCI America™

$135.97 - $391.13

Chemical Identifiers

CAS 32133-82-7
Molecular Formula C30H20F12O2S
Molecular Weight (g/mol) 672.529
MDL Number MFCD00010662
InChI Key RMIBJVUYNZSLSD-UHFFFAOYSA-N
Synonym Martin Sulfurane
PubChem CID 3608068
IUPAC Name bis[(1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-yl)oxy]-diphenyl-$l^{4}-sulfane
SMILES C1=CC=C(C=C1)C(C(F)(F)F)(C(F)(F)F)OS(C2=CC=CC=C2)(C3=CC=CC=C3)OC(C4=CC=CC=C4)(C(F)(F)F)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B22001G View Documents TCI America
B22001G
1g Each for $135.97
 
B22005G View Documents TCI America
B22005G
5g Each for $391.13
 
Specifications

Chemical Identifiers

32133-82-7
672.529
RMIBJVUYNZSLSD-UHFFFAOYSA-N
3608068
C1=CC=C(C=C1)C(C(F)(F)F)(C(F)(F)F)OS(C2=CC=CC=C2)(C3=CC=CC=C3)OC(C4=CC=CC=C4)(C(F)(F)F)C(F)(F)F
C30H20F12O2S
MFCD00010662
Martin Sulfurane
bis[(1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-yl)oxy]-diphenyl-$l^{4}-sulfane

Specifications

White-Yellow
1g
C30H20F12O2S
3261
RMIBJVUYNZSLSD-UHFFFAOYSA-N
bis[(1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-yl)oxy]-diphenyl-$l^{4}-sulfane
3608068
Crystal-Powder at 20°C
Bis[alpha,alpha-bis(trifluoromethyl)benzenemethanolato]diphenylsulfur
111°C
32133-82-7
MFCD00010662
Martin Sulfurane
C1=CC=C(C=C1)C(C(F)(F)F)(C(F)(F)F)OS(C2=CC=CC=C2)(C3=CC=CC=C3)OC(C4=CC=CC=C4)(C(F)(F)F)C(F)(F)F
672.529
672.53
≥96.0% (T)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer