missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis-Acrylamide Solution, 30%, 29:1, Hoefer™

Manufacturer:  Hoefer, Inc GR329500

 View more versions of this product

Catalog No. HBGR329500



Description

Description

Polyacrylamide gel electrophoresis (PAGE)
Specifications

Specifications

Bis-Acrylamide Solution
110-26-9
C7H10N2O2
C=CC(=O)NCNC(=O)C=C
154.169
Molecular Biology, Ultrapure
29:1
30%
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
N-[(prop-2-enoylamino)methyl]prop-2-enamide
8041
500mL

Chemical Identifiers

110-26-9
154.169
8041
C=CC(=O)NCNC(=O)C=C
C7H10N2O2
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
N-[(prop-2-enoylamino)methyl]prop-2-enamide
SDS
Documents