Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(acetoxydimethyltin)oxide

Chemical Identifiers

Chemical Name or Material Bis(acetoxydimethyltin)oxide
CAS 5926-79-4
Formula Weight 431.61
MDL Number MFCD00070453
UN Number UN3146
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3961003

  SDS
alfa aesar™
3961003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3961006

  SDS
alfa aesar™
3961006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3961014

  SDS
alfa aesar™
3961014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 431.61
CAS 5926-79-4
Health Hazard 1 Danger
Chemical Name or Material Bis(acetoxydimethyltin)oxide
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 240°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
MDL Number MFCD00070453
UN Number UN3146
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Quantity 1g
Sensitivity Moisture Sensitive
Physical Form Powder
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.