Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(acetonitrile)dichloropalladium(II), Pd 40.5%

Olefin isomerization, preparation of acetals and hemiacetal esters

$147.31 - $147.31

Chemical Identifiers

Chemical Name or Material Bis(acetonitrile)dichloropalladium(II)
CAS 14592-56-4
Formula Weight 259.41
MDL Number MFCD00013122
UN Number UN3439
Synonym cis-Dichlorobis(acetonitrile)palladium(II)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1000203

 
alfa aesar™
1000203*
1g Each for $147.31

AA1000206

 
alfa aesar™
1000206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 259.41
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Bis(acetonitrile)dichloropalladium(II)
Physical Form Powder
MDL Number MFCD00013122
CAS 14592-56-4
UN Number UN3439
Synonym cis-Dichlorobis(acetonitrile)palladium(II)
Quantity 1g
Solubility Information Soluble in acetonitrile, acetone, chloroform
Concentration Pd 40.5%
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Odor Odorless
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.