Please login to your online account to display your discounted pricing

Bis(acetato-O)triphenylbismuth +98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC383460050

 View more versions of this product

Catalog No. AC383460050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 558.37
Physical Form Powder
Quantity 5g
Color Off-white
CAS Min % 98.0
CAS Max % 100.0
Infrared Spectrum Authentic
MDL Number MFCD00092697
CAS 7239-60-3
Chemical Name or Material Bis(acetato-O)triphenylbismuth
Packaging Glass bottle
Elemental Analysis 98.0% min.
Molecular Formula C22H21BiO4
Assay Percent Range 98+%

Specifications

Chemical Name or Material Bis(acetato-O)triphenylbismuth
CAS 7239-60-3
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Molecular Formula C22H21BiO4
Formula Weight 558.37
MDL Number MFCD00092697