Please login to your online account to display your discounted pricing

Bis(acetato-O)triphenylbismuth +98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Bis(acetato-O)triphenylbismuth
CAS 7239-60-3
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Molecular Formula C22H21BiO4
Formula Weight 558.37
MDL Number MFCD00092697
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC383460050

  SDS
thermo fisher scientific
AC383460050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 558.37
Physical Form Powder
Quantity 5g
Color Off-white
CAS Min % 98.0
CAS Max % 100.0
Infrared Spectrum Authentic
MDL Number MFCD00092697
CAS 7239-60-3
Chemical Name or Material Bis(acetato-O)triphenylbismuth
Packaging Glass bottle
Elemental Analysis 98.0% min.
Molecular Formula C22H21BiO4
Assay Percent Range 98+%