missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate, Electronic grade, ≥99% (metals basis)

Manufacturer:  Alfa Aesar™ H5518803

 View more versions of this product

Catalog No. AAH5518803



Specifications

Specifications

Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate
White
1g
(metals basis)
MFCD02683472
UEJFJTOGXLEPIV-UHFFFAOYSA-M
bis(4-tert-butylphenyl)iodanium;4-methylbenzenesulfonate
16216932
Powder
Electronic
Light sensitive
237°C to 240°C
131717-99-2
C27H33IO3S
4,4-di-tert-butyldiphenyliodonium p-toluenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)[O-].CC(C)(C)C1=CC=C(C=C1)[I+]C2=CC=C(C=C2)C(C)(C)C
564.522
564.52
≥99%

Chemical Identifiers

131717-99-2
564.522
UEJFJTOGXLEPIV-UHFFFAOYSA-M
16216932
CC1=CC=C(C=C1)S(=O)(=O)[O-].CC(C)(C)C1=CC=C(C=C1)[I+]C2=CC=C(C=C2)C(C)(C)C
C27H33IO3S
MFCD02683472
4,4-di-tert-butyldiphenyliodonium p-toluenesulfonate
bis(4-tert-butylphenyl)iodanium;4-methylbenzenesulfonate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents