Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(4-nitrophenyl) sulfone, 95%

$69.46 - $256.15

Chemical Identifiers

Chemical Name or Material Bis(4-nitrophenyl) sulfone
CAS 1156-50-9
Molecular Formula C12H8N2O6S
Formula Weight 308.27
MDL Number MFCD00010447
Synonym 4-Nitrophenyl sulfone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2139706

  SDS
alfa aesar™
B2139706
5g Each for $69.46

AAB2139714

  SDS
alfa aesar™
B2139714
25g Each for $256.15
Description & Specifications

Specifications

Formula Weight 308.27
Chemical Name or Material Bis(4-nitrophenyl) sulfone
TSCA TSCA
Assay 95%
Molecular Formula C12H8N2O6S
MDL Number MFCD00010447
CAS 1156-50-9
Synonym 4-Nitrophenyl sulfone
Melting Point 252° to 255°C
Quantity 5g
Health Hazard 1 Warning
Odor Odorless
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number WR4700000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.