missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(4-methoxyphenyl) Selenoxide, TCI America™

$207.78 - $661.55

Chemical Identifiers

CAS 25862-14-0
Molecular Formula C14H14O3Se
Molecular Weight (g/mol) 309.234
MDL Number MFCD00059043
InChI Key SXEWETDJNQVWQY-UHFFFAOYSA-N
PubChem CID 11141388
IUPAC Name 1-methoxy-4-(4-methoxyphenyl)seleninylbenzene
SMILES COC1=CC=C(C=C1)[Se](=O)C2=CC=C(C=C2)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B13751G View Documents TCI America
B13751G
1g Each for $207.78
 
B13755G View Documents TCI America
B13755G
5g Each for $661.55
 
Specifications

Chemical Identifiers

25862-14-0
309.234
SXEWETDJNQVWQY-UHFFFAOYSA-N
1-methoxy-4-(4-methoxyphenyl)seleninylbenzene
C14H14O3Se
MFCD00059043
11141388
COC1=CC=C(C=C1)[Se](=O)C2=CC=C(C=C2)OC

Specifications

White-Yellow
25862-14-0
MFCD00059043
SXEWETDJNQVWQY-UHFFFAOYSA-N
1-methoxy-4-(4-methoxyphenyl)seleninylbenzene
11141388
Crystal-Powder at 20°C
Bis(4-methoxyphenyl) Selenoxide
1g
C14H14O3Se
3283
COC1=CC=C(C=C1)[Se](=O)C2=CC=C(C=C2)OC
309.234
309.22
≥93.0% (HPLC)
Safety and Handling

Safety and Handling

TSCA : No