missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(4-methoxyphenyl)phenylphosphine, 95%

$50.86 - $209.43

Chemical Identifiers

CAS 14180-51-9
Molecular Formula C20H19O2P
Formula Weight 322.34
MDL Number MFCD00048993
Chemical Name or Material Bis(4-methoxyphenyl)phenylphosphine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2535606 SDS Alfa Aesar™
B2535606
5g Each for $50.86
AAB2535614 SDS Alfa Aesar™
B2535614
25g Each for $209.43
Specifications Description & Specifications

Chemical Identifiers

14180-51-9
322.34
Bis(4-methoxyphenyl)phenylphosphine
C20H19O2P
MFCD00048993

Specifications

14180-51-9
322.34
88° to 90°C
Bis(4-methoxyphenyl)phenylphosphine
C20H19O2P
MFCD00048993
5g
95%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning