Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(4-methoxyphenyl)dimethylsilane, 97%

$166.61 - $166.61

Chemical Identifiers

Chemical Name or Material Bis(4-methoxyphenyl)dimethylsilane
CAS 69983-36-4
Molecular Formula C16H20O2Si
Formula Weight 272.41g/mol
MDL Number MFCD09037964
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2804506

 
alfa aesar™
H28045-06
5g Each for $166.61
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material Bis(4-methoxyphenyl)dimethylsilane
CAS 69983-36-4
Assay 97%
Molecular Formula C16H20O2Si
Formula Weight 272.41g/mol
MDL Number MFCD09037964