Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(4-methoxyphenyl) carbonate, 97%

$74.51 - $74.51

Chemical Identifiers

Chemical Name or Material Bis(4-methoxyphenyl) carbonate
CAS 5676-71-1
Molecular Formula C15H14O5
Formula Weight 274.28
MDL Number MFCD00798557
Synonym 4-Methoxyphenyl carbonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1870209

  SDS
alfa aesar™
A1870209
10g Each for $74.51
Description & Specifications

Specifications

Formula Weight 274.28
Chemical Name or Material Bis(4-methoxyphenyl) carbonate
Assay 97%
Molecular Formula C15H14O5
MDL Number MFCD00798557
CAS 5676-71-1
Synonym 4-Methoxyphenyl carbonate
Melting Point 86° to 88°C
Quantity 10g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.