missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(4-methoxybenzyl)amine, 97%

Manufacturer:  Alfa Aesar™ H5518003

 View more versions of this product

Catalog No. AAH5518003



Specifications

Specifications

Bis(4-methoxybenzyl)amine
17061-62-0
MFCD00277836
benzenemethanamine, 4-methoxy-n-4-methoxyphenyl methyl
COC1=CC=C(C=C1)CNCC2=CC=C(C=C2)OC
257.333
257.33
1g
C16H19NO2
UN3259
HBKPDEWGANZHJO-UHFFFAOYSA-N
1-(4-methoxyphenyl)-N-[(4-methoxyphenyl)methyl]methanamine
714952
97%

Chemical Identifiers

17061-62-0
257.333
HBKPDEWGANZHJO-UHFFFAOYSA-N
714952
COC1=CC=C(C=C1)CNCC2=CC=C(C=C2)OC
C16H19NO2
MFCD00277836
benzenemethanamine, 4-methoxy-n-4-methoxyphenyl methyl
1-(4-methoxyphenyl)-N-[(4-methoxyphenyl)methyl]methanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents