missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(4-hydroxyphenyl) sulfone, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1734230
Description
It is a reactant used in epoxy reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a reactant used in epoxy reactions.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from strong oxidizing agents.
Specifications
Bis(4-hydroxyphenyl) sulfone | |
245°C to 250°C | |
C12H10O4S | |
250 g | |
4,4'-sulfonyldiphenol, bisphenol s, bis 4-hydroxyphenyl sulfone, phenol, 4,4'-sulfonylbis, 4,4'-dihydroxydiphenyl sulfone, 4-hydroxyphenyl sulfone, bis p-hydroxyphenyl sulfone, 4,4'-sulfonylbisphenol, diphone c | |
VPWNQTHUCYMVMZ-UHFFFAOYSA-N | |
4-(4-hydroxyphenyl)sulfonylphenol | |
6626 | |
250.28 |
80-09-1 | |
1.366 | |
MFCD00002350 | |
2052954 | |
Insoluble in water. | |
OC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(O)C=C1 | |
250.27 | |
CHEBI:34372 | |
99% |
Chemical Identifiers
80-09-1 | |
250.27 | |
VPWNQTHUCYMVMZ-UHFFFAOYSA-N | |
6626 | |
4-(4-hydroxyphenyl)sulfonylphenol |
C12H10O4S | |
MFCD00002350 | |
4,4'-sulfonyldiphenol, bisphenol s, bis 4-hydroxyphenyl sulfone, phenol, 4,4'-sulfonylbis, 4,4'-dihydroxydiphenyl sulfone, 4-hydroxyphenyl sulfone, bis p-hydroxyphenyl sulfone, 4,4'-sulfonylbisphenol, diphone c | |
CHEBI:34372 | |
OC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(O)C=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280i-P305+P351+P338
H319-H500
EINECSNumber : 201-250-5
RTECSNumber : SM8925000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only