missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(4-hydroxy-3,5-dimethylphenyl) Sulfone, TCI America™

$55.00 - $418.63

Chemical Identifiers

CAS 13288-70-5
Molecular Formula C16H18O4S
Molecular Weight (g/mol) 306.376
MDL Number MFCD00036032
InChI Key SUCTVKDVODFXFX-UHFFFAOYSA-N
Synonym Bis(3,5-dimethyl-4-hydroxyphenyl) Sulfone
PubChem CID 83310
IUPAC Name 4-(4-hydroxy-3,5-dimethylphenyl)sulfonyl-2,6-dimethylphenol
SMILES CC1=CC(=CC(=C1O)C)S(=O)(=O)C2=CC(=C(C(=C2)C)O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B157725G View Documents TCI America
B157725G
25g Each for $55.00
 
B1577500G View Documents TCI America
B1577500G
500g Each for $418.63
 
Specifications

Chemical Identifiers

13288-70-5
306.376
SUCTVKDVODFXFX-UHFFFAOYSA-N
83310
CC1=CC(=CC(=C1O)C)S(=O)(=O)C2=CC(=C(C(=C2)C)O)C
C16H18O4S
MFCD00036032
Bis(3,5-dimethyl-4-hydroxyphenyl) Sulfone
4-(4-hydroxy-3,5-dimethylphenyl)sulfonyl-2,6-dimethylphenol

Specifications

White-Yellow
25g
C16H18O4S
Bis(3,5-dimethyl-4-hydroxyphenyl) Sulfone
CC1=CC(=CC(=C1O)C)S(=O)(=O)C2=CC(=C(C(=C2)C)O)C
306.376
306.38
≥98.0% (HPLC,T)
297°C
13288-70-5
MFCD00036032
SUCTVKDVODFXFX-UHFFFAOYSA-N
4-(4-hydroxy-3,5-dimethylphenyl)sulfonyl-2,6-dimethylphenol
83310
Crystal-Powder at 20°C
Bis(4-hydroxy-3,5-dimethylphenyl) Sulfone
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2169

TSCA : Yes