Please login to your online account to display your discounted pricing

Bis-(4-hydroxy-1-naphthyl)phenylmethanol, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  301640250

 View more versions of this product

Catalog No. AC301640250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 392.46
Physical Form Powder
Quantity 25g
Color Red-brown
EINECS Number 230-116-9
Beilstein 06, IV, 7666
Infrared Spectrum Authentic
Specific Extinction 350 min. (648nm, 0.01N NaOH)
MDL Number MFCD00003979
Molecular Formula C27H20O3
CAS 6948-88-5
Chemical Name or Material Bis-(4-hydroxy-1-naphthyl)phenylmethanol
Packaging Glass bottle
Synonym α-Hydroxy-α-(4-hydroxy-1-naphthalenyl)-α-phenyl-1-naphthalenemethanol
Visual Transition Interval from pH 8.2 (orange) to pH 10.0 (bluish-green)

Specifications

Chemical Name or Material Bis-(4-hydroxy-1-naphthyl)phenylmethanol
CAS 6948-88-5
Molecular Formula C27H20O3
Formula Weight 392.46
MDL Number MFCD00003979
Synonym α-Hydroxy-α-(4-hydroxy-1-naphthalenyl)-α-phenyl-1-naphthalenemethanol