missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis-(4-hydroxy-1-naphthyl)phenylmethanol, ACROS Organics™

$65.31 - $65.31

Chemical Identifiers

CAS 6948-88-5
Molecular Formula C27H20O3
Molecular Weight (g/mol) 392.454
MDL Number MFCD00003979
InChI Key OUYLDJXFDLBCTQ-UHFFFAOYSA-N
Synonym 4,4'-hydroxy phenyl methylene bis naphthalen-1-ol, 4,4'-hydroxy phenyl methylene di 1-naphthol, 4,4-alpha-hydroxybenzylidene di-1-naphthol, 4,4-hydroxy phenyl methylene bis naphthalen-1-ol, 4-hydroxy 4-hydroxynaphthalen-1-yl benzyl naphthalen-1-ol, 4-hydroxy-4-hydroxynaphthalen-1-yl-phenyl-methyl naphthalen-1-ol, 4-hydroxy-alpha-4-hydroxynaphthyl-alpha-phenylnaphthalene-1-methanol, bis 4-hydroxy-1-naphthyl phenylmethanol, bis 4-hydroxynaphthyl phenylmethan-1-ol, bis-4-hydroxy-1 naphthyl-phenyl-methanol
PubChem CID 81382
IUPAC Name 4-[hydroxy-(4-hydroxynaphthalen-1-yl)-phenylmethyl]naphthalen-1-ol
SMILES C1=CC=C(C=C1)C(C2=CC=C(C3=CC=CC=C32)O)(C4=CC=C(C5=CC=CC=C54)O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC301640050
SDS View Documents
Thermo Fisher Scientific
301640050
5g Glass Bottle
Each for $65.31
 
Description

Description

Specifications

Chemical Identifiers

6948-88-5
392.454
OUYLDJXFDLBCTQ-UHFFFAOYSA-N
81382
C1=CC=C(C=C1)C(C2=CC=C(C3=CC=CC=C32)O)(C4=CC=C(C5=CC=CC=C54)O)O
C27H20O3
MFCD00003979
4,4'-hydroxy phenyl methylene bis naphthalen-1-ol, 4,4'-hydroxy phenyl methylene di 1-naphthol, 4,4-alpha-hydroxybenzylidene di-1-naphthol, 4,4-hydroxy phenyl methylene bis naphthalen-1-ol, 4-hydroxy 4-hydroxynaphthalen-1-yl benzyl naphthalen-1-ol, 4-hydroxy-4-hydroxynaphthalen-1-yl-phenyl-methyl naphthalen-1-ol, 4-hydroxy-alpha-4-hydroxynaphthyl-alpha-phenylnaphthalene-1-methanol, bis 4-hydroxy-1-naphthyl phenylmethanol, bis 4-hydroxynaphthyl phenylmethan-1-ol, bis-4-hydroxy-1 naphthyl-phenyl-methanol
4-[hydroxy-(4-hydroxynaphthalen-1-yl)-phenylmethyl]naphthalen-1-ol

Specifications

Authentic
Red to Brown
5g
C27H20O3
06, IV, 7666
OUYLDJXFDLBCTQ-UHFFFAOYSA-N
4-[hydroxy-(4-hydroxynaphthalen-1-yl)-phenylmethyl]naphthalen-1-ol
81382
Powder
Glass Bottle
121.0°C to 125.0°C
6948-88-5
MFCD00003979
4,4'-hydroxy phenyl methylene bis naphthalen-1-ol, 4,4'-hydroxy phenyl methylene di 1-naphthol, 4,4-alpha-hydroxybenzylidene di-1-naphthol, 4,4-hydroxy phenyl methylene bis naphthalen-1-ol, 4-hydroxy 4-hydroxynaphthalen-1-yl benzyl naphthalen-1-ol, 4-hydroxy-4-hydroxynaphthalen-1-yl-phenyl-methyl naphthalen-1-ol, 4-hydroxy-alpha-4-hydroxynaphthyl-alpha-phenylnaphthalene-1-methanol, bis 4-hydroxy-1-naphthyl phenylmethanol, bis 4-hydroxynaphthyl phenylmethan-1-ol, bis-4-hydroxy-1 naphthyl-phenyl-methanol
C1=CC=C(C=C1)C(C2=CC=C(C3=CC=CC=C32)O)(C4=CC=C(C5=CC=CC=C54)O)O
392.454
392.46
Bis-(4-hydroxy-1-naphthyl)phenylmethanol
Safety and Handling

Safety and Handling

EINECSNumber : 230-116-9

SDS