Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(4-fluorophenyl)methane, 98+%

$108.34 - $108.34

Chemical Identifiers

Chemical Name or Material Bis(4-fluorophenyl)methane
CAS 457-68-1
Molecular Formula C13H10F2
Formula Weight 204.22
MDL Number MFCD00000360
Beilstein 1956387
Synonym 4,4'-Difluorodiphenylmethane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2164522

  SDS
alfa aesar™
B2164522
100g Each for $108.34
Description & Specifications

Specifications

Formula Weight 204.22
Chemical Name or Material Bis(4-fluorophenyl)methane
Quantity 100g
Beilstein 1956387
Assay ≥98%
MDL Number MFCD00000360
Melting Point 27° to 30°C
Synonym 4,4'-Difluorodiphenylmethane
Boiling Point 265°C
Molecular Formula C13H10F2
Flash Point 134°C (273°F)
CAS 457-68-1

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.