missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(4-fluorophenyl) Ether, TCI America™

$33.61 - $88.61

Chemical Identifiers

CAS 330-93-8
Molecular Formula C12H8F2O
Molecular Weight (g/mol) 206.192
InChI Key UUKHFGSOCZLVJO-UHFFFAOYSA-N
Synonym bis 4-fluorophenyl ether, bis 4-fluorophenyl ether, 4,4'-oxybis fluorobenzene, 4-fluorophenyl ether, 4,4'-difluorodiphenyl ether, 1-fluoro-4-4-fluorophenoxy benzene, 1,1'-oxybis 4-fluorobenzene, unii-9r9o553twz, p-fluorophenyl ether, benzene, 1,1'-oxybis 4-fluoro
PubChem CID 67615
IUPAC Name 1-fluoro-4-(4-fluorophenoxy)benzene
SMILES C1=CC(=CC=C1OC2=CC=C(C=C2)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B26481G View Documents TCI America
B26481G
1g Each for $33.61
 
B26485G View Documents TCI America
B26485G
5g Each for $88.61
 
Specifications

Chemical Identifiers

330-93-8
206.192
bis 4-fluorophenyl ether, bis 4-fluorophenyl ether, 4,4'-oxybis fluorobenzene, 4-fluorophenyl ether, 4,4'-difluorodiphenyl ether, 1-fluoro-4-4-fluorophenoxy benzene, 1,1'-oxybis 4-fluorobenzene, unii-9r9o553twz, p-fluorophenyl ether, benzene, 1,1'-oxybis 4-fluoro
1-fluoro-4-(4-fluorophenoxy)benzene
C12H8F2O
UUKHFGSOCZLVJO-UHFFFAOYSA-N
67615
C1=CC(=CC=C1OC2=CC=C(C=C2)F)F

Specifications

240°C
1g
C12H8F2O
UUKHFGSOCZLVJO-UHFFFAOYSA-N
1-fluoro-4-(4-fluorophenoxy)benzene
67615
Clear Liquid at 20°C
Bis(4-fluorophenyl) Ether
Yellow
330-93-8
bis 4-fluorophenyl ether, bis 4-fluorophenyl ether, 4,4'-oxybis fluorobenzene, 4-fluorophenyl ether, 4,4'-difluorodiphenyl ether, 1-fluoro-4-4-fluorophenoxy benzene, 1,1'-oxybis 4-fluorobenzene, unii-9r9o553twz, p-fluorophenyl ether, benzene, 1,1'-oxybis 4-fluoro
C1=CC(=CC=C1OC2=CC=C(C=C2)F)F
206.192
206.19
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No