missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(4-fluorobenzyl)amine, 97%

Manufacturer:  Alfa Aesar™ H5646703

 View more versions of this product

Catalog No. AAH5646703



Specifications

Specifications

Bis(4-fluorobenzyl)amine
134227-41-1
MFCD05155916
bis 4-fluorobenzyl amine, bis 4-fluorophenyl methyl amine, bis p-fluorobenzyl amine, di-4-fluorobenzyl amine, bis-4-fluoro-benzyl-amine, n,n-bis 4-fluorobenzyl amine, 1-4-fluorophenyl-n-4-fluorophenyl methyl methanamine
C1=CC(=CC=C1CNCC2=CC=C(C=C2)F)F
233.262
233.26
1g
C14H13F2N
UN3259
YKFNPVWWPPJNAQ-UHFFFAOYSA-N
1-(4-fluorophenyl)-N-[(4-fluorophenyl)methyl]methanamine
834401
97%

Chemical Identifiers

134227-41-1
233.262
YKFNPVWWPPJNAQ-UHFFFAOYSA-N
834401
C1=CC(=CC=C1CNCC2=CC=C(C=C2)F)F
C14H13F2N
MFCD05155916
bis 4-fluorobenzyl amine, bis 4-fluorophenyl methyl amine, bis p-fluorobenzyl amine, di-4-fluorobenzyl amine, bis-4-fluoro-benzyl-amine, n,n-bis 4-fluorobenzyl amine, 1-4-fluorophenyl-n-4-fluorophenyl methyl methanamine
1-(4-fluorophenyl)-N-[(4-fluorophenyl)methyl]methanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

Documents