Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(4-fluoro-3-nitrophenyl) sulfone, 97%

$87.74 - $87.74

Chemical Identifiers

Chemical Name or Material Bis(4-fluoro-3-nitrophenyl) sulfone
CAS 312-30-1
Molecular Formula C12H6F2N2O6S
Formula Weight 344.25
MDL Number MFCD00007057
Beilstein 2486439
Synonym 4,4'-Difluoro-3,3'-dinitrodiphenyl sulfone; 4-Fluoro-3-nitrophenyl sulfone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1674206

 
alfa aesar™
L1674206
5g Each for $87.74
Description & Specifications

Specifications

Formula Weight 344.25
Chemical Name or Material Bis(4-fluoro-3-nitrophenyl) sulfone
CAS 312-30-1
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay 97%
Molecular Formula C12H6F2N2O6S
MDL Number MFCD00007057
Synonym 4,4'-Difluoro-3,3'-dinitrodiphenyl sulfone; 4-Fluoro-3-nitrophenyl sulfone
Health Hazard 1 Warning
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 2486439
Melting Point 191° to 194°C
RTECS Number WR4300000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.