missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(4-diethylaminophenyl)methanol, TCI America™

$93.20 - $484.31

Chemical Identifiers

CAS 134-91-8
Molecular Formula C21H30N2O
Molecular Weight (g/mol) 326.484
MDL Number MFCD00035979
InChI Key WCIQBUVXZZYFJP-UHFFFAOYSA-N
Synonym 4,4′-Bis(diethylamino)benzhydrol
PubChem CID 67257
IUPAC Name bis[4-(diethylamino)phenyl]methanol
SMILES CCN(CC)C1=CC=C(C=C1)C(C2=CC=C(C=C2)N(CC)CC)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B172510G View Documents TCI America
B172510G
10g Each for $484.31
 
B17255G View Documents TCI America
B17255G
5g Each for $328.48
 
B17251G View Documents TCI America
B17251G
1g Each for $93.20
 
Specifications

Chemical Identifiers

134-91-8
326.484
WCIQBUVXZZYFJP-UHFFFAOYSA-N
67257
CCN(CC)C1=CC=C(C=C1)C(C2=CC=C(C=C2)N(CC)CC)O
C21H30N2O
MFCD00035979
4,4′-Bis(diethylamino)benzhydrol
bis[4-(diethylamino)phenyl]methanol

Specifications

White
10g
C21H30N2O
4,4′C-Bis(diethylamino)benzhydrol
CCN(CC)C1=CC=C(C=C1)C(C2=CC=C(C=C2)N(CC)CC)O
326.484
326.48
≥98.0% (T)
82°C
134-91-8
MFCD00035979
WCIQBUVXZZYFJP-UHFFFAOYSA-N
bis[4-(diethylamino)phenyl]methanol
67257
Crystal-Powder at 20°C
Bis(4-diethylaminophenyl)methanol
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0883

TSCA : No

Recommended Storage : Refrigerator