Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(4-chlorophenylthio)methane, 99%

$34.32 - $113.59

Chemical Identifiers

Chemical Name or Material Bis(4-chlorophenylthio)methane
CAS 2393-97-7
Molecular Formula C13H10Cl2S2
Formula Weight 301.26
MDL Number MFCD00041407
Beilstein 2216758
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2201803

  SDS
alfa aesar™
B2201803
1g Each for $34.32

AAB2201806

  SDS
alfa aesar™
B2201806
5g Each for $113.59
Description & Specifications

Specifications

Formula Weight 301.26
Chemical Name or Material Bis(4-chlorophenylthio)methane
Assay 99%
Molecular Formula C13H10Cl2S2
MDL Number MFCD00041407
CAS 2393-97-7
Beilstein 2216758
Melting Point 45° to 48°C
Boiling Point 193° to 195°C (2mmHg)
Quantity 1g