Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(4-chlorophenyl) sulfoxide, 98%

$30.20 - $30.20

Chemical Identifiers

Chemical Name or Material Bis (4-chlorophenyl) sulfoxide
CAS 3085-42-5
Molecular Formula C12H8Cl2OS
Formula Weight 272.17g/mol
MDL Number MFCD00000618
Synonym 4-Chlorophenyl sulfoxide; 4,4'-Dichlorodiphenyl sulfoxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1100406

 
alfa aesar™
A11004-06
5g Each for $30.20
Description & Specifications

Specifications

Quantity 5g
Melting Point 142° to 144°C
Quantity 5g
Chemical Name or Material Bis (4-chlorophenyl) sulfoxide
CAS 3085-42-5
Assay 98%
Molecular Formula C12H8Cl2OS
Formula Weight 272.17g/mol
MDL Number MFCD00000618
Synonym 4-Chlorophenyl sulfoxide; 4,4'-Dichlorodiphenyl sulfoxide
RTECS Number WS2275000