missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(4-chlorophenyl) disulfide, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2560414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bis(4-chlorophenyl) disulfide | |
1142-19-4 | |
C12H8Cl2S2 | |
1644663 | |
ZIXXRXGPBFMPFD-UHFFFAOYSA-N | |
1-chloro-4-[(4-chlorophenyl)disulfanyl]benzene | |
14360 | |
≥98% |
71°C to 74°C | |
25 g | |
MFCD00013642 | |
4,4'-dichlorodiphenyl disulfide, bis 4-chlorophenyl disulfide, disulfide, bis 4-chlorophenyl, ddds, ddds pesticide, disulfide, bis p-chlorophenyl, p-chlorophenyl disulfide, 4-chlorophenyl disulfide, bis p-chlorophenyl disulfide, 4,4'-dichloro diphenyl disulfide | |
C1=CC(=CC=C1SSC2=CC=C(C=C2)Cl)Cl | |
287.216 | |
287.23 |
Chemical Identifiers
1142-19-4 | |
287.216 | |
ZIXXRXGPBFMPFD-UHFFFAOYSA-N | |
14360 | |
C1=CC(=CC=C1SSC2=CC=C(C=C2)Cl)Cl |
C12H8Cl2S2 | |
MFCD00013642 | |
4,4'-dichlorodiphenyl disulfide, bis 4-chlorophenyl disulfide, disulfide, bis 4-chlorophenyl, ddds, ddds pesticide, disulfide, bis p-chlorophenyl, p-chlorophenyl disulfide, 4-chlorophenyl disulfide, bis p-chlorophenyl disulfide, 4,4'-dichloro diphenyl disulfide | |
1-chloro-4-[(4-chlorophenyl)disulfanyl]benzene |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-531-2
RTECSNumber : JO0766000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only