missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(4-bromophenylboronic Acid) scyllo-Inositol Complex Dipotassium Tetrahydrate, TCI America™

$85.56 - $253.61

Chemical Identifiers

CAS 1537876-29-1
Molecular Formula C18H14B2Br2K2O6
Molecular Weight (g/mol) 585.929
InChI Key KFMXNLSRMBTKTI-UHFFFAOYSA-N
PubChem CID 91972154
SMILES [B-]12(OC3C4C(O1)C5C(C3O[B-](O4)(O5)C6=CC=C(C=C6)Br)O2)C7=CC=C(C=C7)Br.[K+].[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B4524200MG View Documents TCI America
B4524200MG
200mg Each for $85.56
 
B45241G View Documents TCI America
B45241G
1g Each for $253.61
 
Specifications

Chemical Identifiers

1537876-29-1
585.929
91972154
C18H14B2Br2K2O6
KFMXNLSRMBTKTI-UHFFFAOYSA-N
[B-]12(OC3C4C(O1)C5C(C3O[B-](O4)(O5)C6=CC=C(C=C6)Br)O2)C7=CC=C(C=C7)Br.[K+].[K+]

Specifications

White-Yellow
1537876-29-1
KFMXNLSRMBTKTI-UHFFFAOYSA-N
585.929
585.93
≥98.0% (HPLC)
200mg
C18H14B2Br2K2O6
[B-]12(OC3C4C(O1)C5C(C3O[B-](O4)(O5)C6=CC=C(C=C6)Br)O2)C7=CC=C(C=C7)Br.[K+].[K+]
91972154
Crystal-Powder at 20°C
Bis(4-bromophenylboronic Acid) scyllo-Inositol Complex Dipotassium Tetrahydrate
Safety and Handling

Safety and Handling

TSCA : No