missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(4-bromophenyl)phenylphosphine Oxide, TCI America™

$85.56 - $271.95

Chemical Identifiers

CAS 93869-52-4
Molecular Formula C18H13Br2OP
Molecular Weight (g/mol) 436.083
InChI Key URGNHDJCYWEAKG-UHFFFAOYSA-N
PubChem CID 15469924
IUPAC Name 1-bromo-4-[(4-bromophenyl)-phenylphosphoryl]benzene
SMILES C1=CC=C(C=C1)P(=O)(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B55321G View Documents TCI America
B55321G
1g Each for $271.95
 
B5532200MG View Documents TCI America
B5532200MG
200mg Each for $85.56
 
Specifications

Chemical Identifiers

93869-52-4
436.083
15469924
C1=CC=C(C=C1)P(=O)(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br
C18H13Br2OP
URGNHDJCYWEAKG-UHFFFAOYSA-N
1-bromo-4-[(4-bromophenyl)-phenylphosphoryl]benzene

Specifications

280°C
107°C
93869-52-4
URGNHDJCYWEAKG-UHFFFAOYSA-N
1-bromo-4-[(4-bromophenyl)-phenylphosphoryl]benzene
15469924
Crystal-Powder at 20°C
Bis(4-bromophenyl)phenylphosphine Oxide
White-Yellow
1g
C18H13Br2OP
C1=CC=C(C=C1)P(=O)(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br
436.083
436.08
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No