Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(3-fluorophenyl) disulfide, 97%

$98.66 - $380.20

Chemical Identifiers

Chemical Name or Material Bis(3-fluorophenyl) disulfide
CAS 63930-17-6
Molecular Formula C12H8F2S2
Formula Weight 254.31
MDL Number MFCD01318104
Beilstein 2530329
Synonym 3,3'-Difluorodiphenyl disulfide; 3-Fluorophenyl disulfide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1718603

  SDS
alfa aesar™
L1718603
1g Each for $98.66

AAL1718606

  SDS
alfa aesar™
L1718606
5g Each for $380.20
Description & Specifications

Specifications

Formula Weight 254.31
Chemical Name or Material Bis(3-fluorophenyl) disulfide
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Molecular Formula C12H8F2S2
MDL Number MFCD01318104
Beilstein 2530329
Synonym 3,3'-Difluorodiphenyl disulfide; 3-Fluorophenyl disulfide
Health Hazard 1 Warning
Boiling Point 308°C
Quantity 1g
Flash Point >110°C (230°F)
CAS 63930-17-6
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.