Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(3-chlorobenzyl)amine, 97%

$1,367.35 - $1,367.35

Chemical Identifiers

Chemical Name or Material Bis(3-chlorobenzyl)amine
CAS 129041-31-2
Molecular Formula C14H13Cl2N
Formula Weight 266.17g/mol
MDL Number MFCD00671501
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2508022

 
alfa aesar™
B25080-22
100g Each for $1,367.35
Description & Specifications

Specifications

Quantity 100g
Chemical Name or Material Bis(3-chlorobenzyl)amine
CAS 129041-31-2
Assay 97%
Molecular Formula C14H13Cl2N
Formula Weight 266.17g/mol
MDL Number MFCD00671501
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.