Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(3-aminopropyl)phenylphosphine

Chemical Identifiers

Chemical Name or Material Bis(3-aminopropyl)phenylphosphine
CAS 6775-01-5
Formula Weight 224.29
MDL Number MFCD00014834
UN Number UN3267
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1042704

 
alfa aesar™
1042704
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1042709

 
alfa aesar™
1042709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 224.29
Chemical Name or Material Bis(3-aminopropyl)phenylphosphine
Refractive Index 1.5728
Physical Form Liquid
Density 1.03g/mL
MDL Number MFCD00014834
CAS 6775-01-5
UN Number UN3267
Boiling Point 144°C (1mmHg)
Quantity 2g
Sensitivity Air Sensitive
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H335-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H335-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes serious eye irritation.