missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(3,5-dimethylphenyl)phosphine oxide, 98%

Manufacturer:  Alfa Aesar™ H6132003

 View more versions of this product

Catalog No. AAH6132003



Specifications

Specifications

Bis(3,5-dimethylphenyl)phosphine oxide
82°C to 84°C
187344-92-9
MFCD08458884
LMXRTXPFJNGAAX-UHFFFAOYSA-N
bis(3,5-dimethylphenyl)-oxophosphanium
12022244
Powder
White
1g
C16H18OP+
1-3,5-dimethylphenyl phosphoryl-3,5-dimethylbenzene
CC1=CC(=CC(=C1)[P+](=O)C2=CC(=CC(=C2)C)C)C
257.293
258.3
98%

Chemical Identifiers

187344-92-9
257.293
LMXRTXPFJNGAAX-UHFFFAOYSA-N
12022244
CC1=CC(=CC(=C1)[P+](=O)C2=CC(=CC(=C2)C)C)C
C16H18OP+
MFCD08458884
1-3,5-dimethylphenyl phosphoryl-3,5-dimethylbenzene
bis(3,5-dimethylphenyl)-oxophosphanium
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents