missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(3,5-dimethylphenyl)phosphine oxide, 98%

$44.20 - $175.19

Chemical Identifiers

CAS 187344-92-9
Molecular Formula C16H18OP+
Molecular Weight (g/mol) 257.293
MDL Number MFCD08458884
InChI Key LMXRTXPFJNGAAX-UHFFFAOYSA-N
Synonym 1-3,5-dimethylphenyl phosphoryl-3,5-dimethylbenzene, 1-3,5-dimethylphenylphosphoroso-3,5-dimethylbenzene, 3-4-nitrophenyl-beta-alanine, awaysumoanjulo-uhfffaoysa-n, bis 3,5-dimethylphenyl phosphi, bis 3,5-dimethylphenyl phosphine oxide, ksc496c0h, phosphine oxide, bis 3,5-dimethylphenyl, phosphine oxide,bis 3,5-dimethylphenyl, xyl 2p o h
PubChem CID 12022244
IUPAC Name bis(3,5-dimethylphenyl)-oxophosphanium
SMILES CC1=CC(=CC(=C1)[P+](=O)C2=CC(=CC(=C2)C)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6132003
View Documents
Alfa Aesar™
H6132003
1g Each for $44.20
 
AAH6132006
View Documents
Alfa Aesar™
H6132006
5g Each for $175.19
 
Specifications

Chemical Identifiers

187344-92-9
257.293
LMXRTXPFJNGAAX-UHFFFAOYSA-N
12022244
CC1=CC(=CC(=C1)[P+](=O)C2=CC(=CC(=C2)C)C)C
C16H18OP+
MFCD08458884
1-3,5-dimethylphenyl phosphoryl-3,5-dimethylbenzene, 1-3,5-dimethylphenylphosphoroso-3,5-dimethylbenzene, 3-4-nitrophenyl-beta-alanine, awaysumoanjulo-uhfffaoysa-n, bis 3,5-dimethylphenyl phosphi, bis 3,5-dimethylphenyl phosphine oxide, ksc496c0h, phosphine oxide, bis 3,5-dimethylphenyl, phosphine oxide,bis 3,5-dimethylphenyl, xyl 2p o h
bis(3,5-dimethylphenyl)-oxophosphanium

Specifications

White
1g
C16H18OP+
1-3,5-dimethylphenyl phosphoryl-3,5-dimethylbenzene, 1-3,5-dimethylphenylphosphoroso-3,5-dimethylbenzene, 3-4-nitrophenyl-beta-alanine, awaysumoanjulo-uhfffaoysa-n, bis 3,5-dimethylphenyl phosphi, bis 3,5-dimethylphenyl phosphine oxide, ksc496c0h, phosphine oxide, bis 3,5-dimethylphenyl, phosphine oxide,bis 3,5-dimethylphenyl, xyl 2p o h
CC1=CC(=CC(=C1)[P+](=O)C2=CC(=CC(=C2)C)C)C
257.293
258.3
98%
82°C to 84°C
187344-92-9
MFCD08458884
LMXRTXPFJNGAAX-UHFFFAOYSA-N
bis(3,5-dimethylphenyl)-oxophosphanium
12022244
Powder
Bis(3,5-dimethylphenyl)phosphine oxide
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures