Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(3,5-di-tert-butyl-4-methoxyphenyl)chlorophosphine, 94%

Chemical Identifiers

Chemical Name or Material Bis(3,5-di-tert-butyl-4-methoxyphenyl)chlorophosphine
CAS 212713-08-1
Molecular Formula C30H46ClO2P
Formula Weight 505.13
MDL Number MFCD08458881
UN Number UN3261
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2588203

  SDS
alfa aesar™
H2588203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2588206

  SDS
alfa aesar™
H2588206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 505.13
UN Number UN3261
Chemical Name or Material Bis(3,5-di-tert-butyl-4-methoxyphenyl)chlorophosphine
Molecular Formula C30H46ClO2P
Quantity 1g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD08458881
CAS 212713-08-1
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Assay 94%

Applications
It is used as a reactant for asymmetric hydrogenation with iridium catalysts, chemo selective reaction with deprotonated 2-hydoxy-3,3,N-trimethylbutanamide. It is also used as a reactant for synthesis of phosphine- containing amino acids and peptide-based catalyst ligands, nonracemic phosphine-containing amino acids for palladium- catalyzed asymmetric allylic substitution reactions.

Solubility
Reacts with water.

Notes
Air & moisture sensitive. Materials to avoid is oxidizing agents, bases.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.