missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis[(2R,5R)-1-(2-[(2R,5R)-2,5-dimethyl-1-phospholanyl]phenyl)-2,5-dimethylphospholane 1-oxide]copper(I) triflate, 97%, Alfa Aesar™

$96.38 - $382.45

Chemical Identifiers

CAS 874013-62-4
Molecular Formula C37H56CuF3O5P4S
Molecular Weight (g/mol) 857.346
MDL Number MFCD17018797
InChI Key JGVWSKIVCRMXGT-YCIFJGFLSA-M
Synonym bis 1-2r,5r-2,5-dimethylphospholanyl-2-2r,5r-2,5-dimethylphospholanyl-oxide benzene cu i trifluoromethanesulfonate
PubChem CID 11707733
IUPAC Name copper(1+);(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethyl-1$l^{5}-phospholane 1-oxide;trifluoromethanesulfonate
SMILES CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.C(F)(F)(F)S(=O)(=O)[O-].[Cu+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63492MC View Documents Alfa Aesar
H63492MC
100mg Each for $96.38
 
AAH63492ME View Documents Alfa Aesar
H63492ME
500mg Each for $382.45
 
Specifications

Chemical Identifiers

874013-62-4
857.346
JGVWSKIVCRMXGT-YCIFJGFLSA-M
11707733
CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.C(F)(F)(F)S(=O)(=O)[O-].[Cu+]
C37H56CuF3O5P4S
MFCD17018797
bis 1-2r,5r-2,5-dimethylphospholanyl-2-2r,5r-2,5-dimethylphospholanyl-oxide benzene cu i trifluoromethanesulfonate
copper(1+);(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethyl-1$l^{5}-phospholane 1-oxide;trifluoromethanesulfonate

Specifications

Air Sensitive
100mg
C37H56CuF3O5P4S
bis 1-2r,5r-2,5-dimethylphospholanyl-2-2r,5r-2,5-dimethylphospholanyl-oxide benzene cu i trifluoromethanesulfonate
CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.C(F)(F)(F)S(=O)(=O)[O-].[Cu+]
857.346
857.34
97%
Brown to White
874013-62-4
MFCD17018797
JGVWSKIVCRMXGT-YCIFJGFLSA-M
copper(1+);(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethyl-1$l^{5}-phospholane 1-oxide;trifluoromethanesulfonate
11707733
Solid
Bis[(2R,5R)-1-(2-[(2R,5R)-2,5-dimethyl-1-phospholanyl]phenyl)-2,5-dimethylphospholane 1-oxide]copper(I) triflate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning