missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis[(2R,5R)-1-(2-[(2R,5R)-2,5-dimethyl-1-phospholanyl]phenyl)-2,5-dimethylphospholane 1-oxide]copper(I) triflate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H63492MC
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bis[(2R,5R)-1-(2-[(2R,5R)-2,5-dimethyl-1-phospholanyl]phenyl)-2,5-dimethylphospholane 1-oxide]copper | |
Brown to White | |
MFCD17018797 | |
Air Sensitive | |
JGVWSKIVCRMXGT-YCIFJGFLSA-M | |
copper(1+);(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethyl-1$l^{5}-phospholane 1-oxide;trifluoromethanesulfonate | |
11707733 | |
97% |
874013-62-4 | |
C37H56CuF3O5P4S | |
100 mg | |
bis 1-2r,5r-2,5-dimethylphospholanyl-2-2r,5r-2,5-dimethylphospholanyl-oxide benzene cu i trifluoromethanesulfonate | |
CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.C(F)(F)(F)S(=O)(=O)[O-].[Cu+] | |
857.346 | |
857.34 | |
Solid |
Chemical Identifiers
874013-62-4 | |
857.346 | |
JGVWSKIVCRMXGT-YCIFJGFLSA-M | |
11707733 | |
CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.C(F)(F)(F)S(=O)(=O)[O-].[Cu+] |
C37H56CuF3O5P4S | |
MFCD17018797 | |
bis 1-2r,5r-2,5-dimethylphospholanyl-2-2r,5r-2,5-dimethylphospholanyl-oxide benzene cu i trifluoromethanesulfonate | |
copper(1+);(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethyl-1$l^{5}-phospholane 1-oxide;trifluoromethanesulfonate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only