Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(2-thiophenemethyl)amine hydrochloride, 99%

$86.66 - $86.66

Chemical Identifiers

Chemical Name or Material Bis(2-thiophenemethyl)amine hydrochloride
CAS 336624-10-3
Molecular Formula C10H11NS2·HCl
Formula Weight 245.8
MDL Number MFCD01812493
Synonym Di-2-thenylamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2229009

  SDS
alfa aesar™
B2229009
10g Each for $86.66
Description & Specifications

Specifications

Formula Weight 245.8
Chemical Name or Material Bis(2-thiophenemethyl)amine hydrochloride
Assay 99%
Molecular Formula C10H11NS2·HCl
MDL Number MFCD01812493
CAS 336624-10-3
Synonym Di-2-thenylamine hydrochloride
Melting Point ≈243°C (dec.)
Quantity 10g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.