Learn More
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride, 97%, Thermo Scientific Chemicals
$100.19 - $327.30
Chemical Identifiers
CAS | 68641-49-6 |
---|---|
Molecular Formula | C6H8ClN2O5P |
Molecular Weight (g/mol) | 254.563 |
MDL Number | MFCD00010077 |
InChI Key | KLDLRDSRCMJKGM-UHFFFAOYSA-N |
Synonym | bis 2-oxo-3-oxazolidinyl phosphinic chloride, bop-cl, bis 2-oxooxazolidin-3-yl phosphinic chloride, ccris 2607, phosphinic chloride, bis 2-oxo-3-oxazolidinyl, bis 2-oxo-3-oxazolidinyl phosphonic chloride, bis 2-oxo-1,3-oxazolidin-3-yl phosphinoyl chloride, bis-2-oxo-3-oxazolidinyl phosphinic chloride, n,n-bis 2-oxo-3-oxazolidinyl phosphinic chloride, bis 2-oxo-3-oxazolidinyl phosphinic chloride bop-cl |
PubChem CID | 152842 |
IUPAC Name | 3-[chloro-(2-oxo-1,3-oxazolidin-3-yl)phosphoryl]-1,3-oxazolidin-2-one |
SMILES | C1COC(=O)N1P(=O)(N2CCOC2=O)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0877506
|
Thermo Scientific Chemicals
L0877506 |
5 g |
Each for $100.19
|
|
|||||
AAL0877514
|
Thermo Scientific Chemicals
L0877514 |
25 g |
Each for $327.30
|
|
|||||
Description
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride was used in the preparation of hexadepsipeptide. Also used as reagent is used for activating the carboxylic group, synthesis of amides, esters and peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBis(2-oxo-3-oxazolidinyl)phosphinic chloride was used in the preparation of hexadepsipeptide. Also used as reagent is used for activating the carboxylic group, synthesis of amides, esters and peptides.
Solubility
Hydrolyzes with water.
Notes
Moisture Sensitive, store away from moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
68641-49-6 | |
254.563 | |
KLDLRDSRCMJKGM-UHFFFAOYSA-N | |
152842 | |
C1COC(=O)N1P(=O)(N2CCOC2=O)Cl |
C6H8ClN2O5P | |
MFCD00010077 | |
bis 2-oxo-3-oxazolidinyl phosphinic chloride, bop-cl, bis 2-oxooxazolidin-3-yl phosphinic chloride, ccris 2607, phosphinic chloride, bis 2-oxo-3-oxazolidinyl, bis 2-oxo-3-oxazolidinyl phosphonic chloride, bis 2-oxo-1,3-oxazolidin-3-yl phosphinoyl chloride, bis-2-oxo-3-oxazolidinyl phosphinic chloride, n,n-bis 2-oxo-3-oxazolidinyl phosphinic chloride, bis 2-oxo-3-oxazolidinyl phosphinic chloride bop-cl | |
3-[chloro-(2-oxo-1,3-oxazolidin-3-yl)phosphoryl]-1,3-oxazolidin-2-one |
Specifications
68641-49-6 | |
C6H8ClN2O5P | |
5 g | |
3654596 | |
bis 2-oxo-3-oxazolidinyl phosphinic chloride, bop-cl, bis 2-oxooxazolidin-3-yl phosphinic chloride, ccris 2607, phosphinic chloride, bis 2-oxo-3-oxazolidinyl, bis 2-oxo-3-oxazolidinyl phosphonic chloride, bis 2-oxo-1,3-oxazolidin-3-yl phosphinoyl chloride, bis-2-oxo-3-oxazolidinyl phosphinic chloride, n,n-bis 2-oxo-3-oxazolidinyl phosphinic chloride, bis 2-oxo-3-oxazolidinyl phosphinic chloride bop-cl | |
KLDLRDSRCMJKGM-UHFFFAOYSA-N | |
3-[chloro-(2-oxo-1,3-oxazolidin-3-yl)phosphoryl]-1,3-oxazolidin-2-one | |
152842 | |
97% |
∽195°C (decomposition) | |
MFCD00010077 | |
UN3261 | |
Moisture sensitive | |
Hydrolyzes with water. | |
C1COC(=O)N1P(=O)(N2CCOC2=O)Cl | |
254.563 | |
254.57 | |
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RTECSNumber : SZ5871000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only