Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(2-nitrophenyl) disulfide, 98%

$57.53 - $186.66

Chemical Identifiers

Chemical Name or Material Bis(2-nitrophenyl) disulfide
CAS 1155-00-6
Molecular Formula C12H8N2O4S2
Formula Weight 308.33g/mol
MDL Number MFCD00007130
Beilstein 1016770
Synonym 2,2'-Dinitrodiphenyl disulfide; 2-Nitrophenyl disulfide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2561514

 
alfa aesar™
B25615-14
25g Each for $57.53

AAB2561522

 
alfa aesar™
B25615-22
100g Each for $186.66
Description & Specifications

Specifications

Melting Point 192° to 196°C
Quantity 25g
Chemical Name or Material Bis(2-nitrophenyl) disulfide
CAS 1155-00-6
Assay 98%
Molecular Formula C12H8N2O4S2
Formula Weight 308.33g/mol
MDL Number MFCD00007130
Beilstein 1016770
Synonym 2,2'-Dinitrodiphenyl disulfide; 2-Nitrophenyl disulfide
TSCA TSCA