missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis-(2-methylallyl)cycloocta-1,5-diene ruthenium(II) complex, 30-32% Ru, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 295782500

 View more versions of this product

Catalog No. AC295782500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Bis-(2-methylallyl)cycloocta-1, 5-diene ruthenium(II) complex
12289-94-0
94% min.
Crystalline Powder
319.44
Beige-brown to brown
250mg
30-32% Ru
Authentic
Glass bottle
C16H26Ru
MFCD00216965
80° to 84°C

Chemical Identifiers

12289-94-0
319.44
Bis-(2-methylallyl)cycloocta-1, 5-diene ruthenium(II) complex, 30-32% Ru
C16H26Ru
MFCD00216965

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MOISTURE SENSITIVE