Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II)

Chemical Identifiers

Chemical Name or Material Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II)
CAS 12289-94-0
Molecular Formula C16H26Ru
Formula Weight 319.45
MDL Number MFCD00216965
UN Number UN3395
Synonym (1,5-Cyclooctadiene)bis(2-methylallyl)ruthenium(II); Ruthenium(II) bis(2-methylallyl) 1,5-cyclooctadiene complex
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2689103

  SDS
alfa aesar™
H2689103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 319.45
MDL Number MFCD00216965
Health Hazard 2 GHS H Statement
H261
In contact with water releases flammable gas.
Health Hazard 1 Warning
Chemical Name or Material Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II)
Health Hazard 3 GHS P Statement
P231+P232-P280-P370+P378c-P402+P404-P501a
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 12289-94-0
UN Number UN3395
Quantity 1g
Molecular Formula C16H26Ru
Synonym (1,5-Cyclooctadiene)bis(2-methylallyl)ruthenium(II); Ruthenium(II) bis(2-methylallyl) 1,5-cyclooctadiene complex
Recommended Storage −30° to −10°C
Melting Point 83° to 86°C
DOT Information Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, WATER-REACTIVE

Applications
Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II) is used as a catalyst and pharmaceutical intermediate. It is also used as a precatalysts for asymmetric and Cross-Coupling Catalysis reactions, and in organic chemical synthesis.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P231+P232-P280-P370+P378c-P402+P404-P501a
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H261
In contact with water releases flammable gas.

−30° to −10°C