Please login to your online account to display your discounted pricing

Bis(2-methyl-3-furyl)disulphide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Bis(2-methyl-3-furyl)disulphide
CAS 28588-75-2
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C10H10O2S2
Formula Weight 226.30g/mol
MDL Number MFCD01074774
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC332690050

  SDS
thermo fisher scientific
332690050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density (20°C) 1.207 to 1.215g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5720 to 1.5830 (20°C, 589nm)
Color Clear yellow to amber or orange
Quantity 5g
Chemical Name or Material Bis(2-methyl-3-furyl)disulphide
CAS 28588-75-2
CAS Min % 97.5
CAS Max % 100.0
Physical Form Clear yellow to amber or orange liquid
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C10H10O2S2
Formula Weight 226.30g/mol
MDL Number MFCD01074774
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.