Please login to your online account to display your discounted pricing

Bis(2-methyl-3-furyl)disulphide, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  332690050

 View more versions of this product

Catalog No. AC332690050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 226.3
Quantity 5g
Color Clear yellow to amber or orange
CAS Max % 100.0
Chemical Name or Material Bis(2-methyl-3-furyl)disulphide
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Refractive Index 1.5720 to 1.5830 (20°C, 589nm)
Molecular Formula C10H10O2S2
MDL Number MFCD01074774
CAS 28588-75-2
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Density (20°C) 1.207 to 1.215g/mL

Specifications

Chemical Name or Material Bis(2-methyl-3-furyl)disulphide
CAS 28588-75-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H10O2S2
Formula Weight 226.3
MDL Number MFCD01074774
Accessories

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents