missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(2-methoxyphenyl)phenylphosphine 98%, ACROS Organics™

Chemical Identifiers

CAS 36802-41-2
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C20H19O2P
Formula Weight 322.34
MDL Number MFCD00061478
Chemical Name or Material Bis(2-methoxyphenyl)phenylphosphine
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC380020010 SDS Thermo Fisher Scientific
AC380020010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

36802-41-2
100.0
C20H19O2P
MFCD00061478
≥97.5%
97.5
98%
322.34
Bis(2-methoxyphenyl)phenylphosphine

Specifications

Crystals or Crystalline Powder
1g
158° to 165°C
Glass bottle
98%
97.5
C20H19O2P
≥97.5%
322.34
White
100.0
Authentic
MFCD00061478
36802-41-2
Bis(2-methoxyphenyl)phenylphosphine

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.