missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ Bis(2-methoxyphenyl) carbonate, 98%
$65.27 - $233.49
Chemical Identifiers
CAS | 553-17-3 |
---|---|
Molecular Formula | C15H14O5 |
Molecular Weight (g/mol) | 274.272 |
MDL Number | MFCD00025723 |
InChI Key | ORUJFMPWKPVXLZ-UHFFFAOYSA-N |
Synonym | bis 2-methoxyphenyl carbonate, bis-o-methoxyphenyl carbonate, carbonic acid bis 2-methoxyphenyl ester, di-o-methoxyphenyl carbonate, duotal, guaiacol carbonate, guaiacol carbonic acid neutral ester, guaiacyl carbonate, tussophob, unii-q71xpq6r29 |
PubChem CID | 11104 |
IUPAC Name | bis(2-methoxyphenyl) carbonate |
SMILES | COC1=CC=CC=C1OC(=O)OC2=CC=CC=C2OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1282506
|
Alfa Aesar™
L1282506 |
5g |
Each for $65.27
|
|
AAL1282514
|
Alfa Aesar™
L1282514 |
25g |
Each for $233.49
|
|
Chemical Identifiers
553-17-3 | |
274.272 | |
ORUJFMPWKPVXLZ-UHFFFAOYSA-N | |
11104 | |
COC1=CC=CC=C1OC(=O)OC2=CC=CC=C2OC |
C15H14O5 | |
MFCD00025723 | |
bis 2-methoxyphenyl carbonate, bis-o-methoxyphenyl carbonate, carbonic acid bis 2-methoxyphenyl ester, di-o-methoxyphenyl carbonate, duotal, guaiacol carbonate, guaiacol carbonic acid neutral ester, guaiacyl carbonate, tussophob, unii-q71xpq6r29 | |
bis(2-methoxyphenyl) carbonate |
Specifications
86°C to 88°C | |
553-17-3 | |
MFCD00025723 | |
bis 2-methoxyphenyl carbonate, bis-o-methoxyphenyl carbonate, carbonic acid bis 2-methoxyphenyl ester, di-o-methoxyphenyl carbonate, duotal, guaiacol carbonate, guaiacol carbonic acid neutral ester, guaiacyl carbonate, tussophob, unii-q71xpq6r29 | |
COC1=CC=CC=C1OC(=O)OC2=CC=CC=C2OC | |
274.272 | |
274.28 | |
Bis(2-methoxyphenyl) carbonate |
5g | |
C15H14O5 | |
2472687 | |
ORUJFMPWKPVXLZ-UHFFFAOYSA-N | |
bis(2-methoxyphenyl) carbonate | |
11104 | |
98% |
Safety and Handling
EINECSNumber : 209-034-2
TSCA : No
Recommended Storage : Ambient temperatures