Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(2-methoxyphenyl)-1,1,2,2-tetramethyldisilane, 95%

$64.80 - $189.34

Chemical Identifiers

Chemical Name or Material Bis(2-methoxyphenyl)-1,1,2,2-tetramethyldisilane
CAS 332343-84-7
Molecular Formula C18H26O2Si2
Formula Weight 330.57
MDL Number MFCD09265162
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2739503

 
alfa aesar™
H2739503
1g Each for $64.80

AAH2739506

 
alfa aesar™
H2739506
5g Each for $189.34
Description & Specifications

Specifications

Formula Weight 330.57
Chemical Name or Material Bis(2-methoxyphenyl)-1,1,2,2-tetramethyldisilane
Assay 95%
Molecular Formula C18H26O2Si2
MDL Number MFCD09265162
CAS 332343-84-7
Boiling Point 139° to 144°C (4mmHg)
Quantity 1g