missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(2-methoxyethyl) phthalate, 96%

Manufacturer:  Alfa Aesar™ 4408930

 View more versions of this product

Catalog No. AA4408930



Specifications

Specifications

Bis (2-methoxyethyl) phthalate
185°C (365°F)
340°C
250g
C14H18O6
2056929
Soluble in water 8500mg/L at 25°C.
COCCOC(=O)C1=CC=CC=C1C(=O)OCCOC
282.292
282.29
96%
1.17 g/mL
1.503
Colorless
117-82-8
MFCD00042842
2-methoxyethyl phthalate
HSUIVCLOAAJSRE-UHFFFAOYSA-N
bis(2-methoxyethyl) benzene-1,2-dicarboxylate
8344
Liquid

Chemical Identifiers

117-82-8
282.292
HSUIVCLOAAJSRE-UHFFFAOYSA-N
8344
COCCOC(=O)C1=CC=CC=C1C(=O)OCCOC
C14H18O6
MFCD00042842
2-methoxyethyl phthalate
bis(2-methoxyethyl) benzene-1,2-dicarboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H360Df

EINECSNumber : 204-212-6

RTECSNumber : TI1400000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents