missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(2-methoxyethyl) Ether (Certified), Fisher Chemical

Manufacturer:  Thermo Scientific O1471500

 View more versions of this product

Catalog No. O1471-500




Specifications

Specifications

2°C max.
0.937g/cm³
Glass Bottle
Undesignated
500mL
≥99 %
CH3OCH2CH2OCH2CH2OCH3
3271
SBZXBUIDTXKZTM-UHFFFAOYSA-N
1-methoxy-2-(2-methoxyethoxy)ethane
8150
134.18
≥99%
Bis(2-methoxyethyl) Ether
Pass Test
162°C
-64°C
111-96-6
C6H14O3
MFCD00008503
1-methoxy-2-2-methoxyethoxy ethane
COCCOCCOC
134.175
CHEBI:46784
Liquid
Certified

Chemical Identifiers

111-96-6
134.175
SBZXBUIDTXKZTM-UHFFFAOYSA-N
8150
1-methoxy-2-(2-methoxyethoxy)ethane
C6H14O3
MFCD00008503
1-methoxy-2-2-methoxyethoxy ethane
CHEBI:46784
COCCOCCOC
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents